Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-Bromotetrahydro-4H-pyran-4-one, 90%

Click to view available options
Quantity:
250mg
1g
98021-79-5
C5H7BrO2
179.013
MFCD13193743
ZGULSQDBOSMIBZ-UHFFFAOYSA-N
3-bromodihydro-2h-pyran-4 3h-one,3-bromotetrahydropyran-4-one,4h-pyran-4-one, 3-bromotetrahydro,3-bromotetrahydro-4h-pyran-4-one,3-bromo-tetrahydropyran-4-one,acmc-20a5hh,3-bromo-tetrahydro-pyran-4-one
22224541
3-bromooxan-4-one
C1COCC(C1=O)Br
98021-79-5
C5H7BrO2
179.013
MFCD13193743
ZGULSQDBOSMIBZ-UHFFFAOYSA-N
3-bromodihydro-2h-pyran-4 3h-one,3-bromotetrahydropyran-4-one,4h-pyran-4-one, 3-bromotetrahydro,3-bromotetrahydro-4h-pyran-4-one,3-bromo-tetrahydropyran-4-one,acmc-20a5hh,3-bromo-tetrahydro-pyran-4-one
22224541
3-bromooxan-4-one
C1COCC(C1=O)Br
Quantity 250mg
Beilstein 109139
Formula Weight 179.01
Physical Form Solid
Percent Purity 90%
Chemical Name or Material 3-Bromotetrahydro-4H-pyran-4-one