Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-Bromothioanisole, 98%

Click to view available options
Quantity:
1g
5g
33733-73-2
C7H7BrS
203.097
MFCD00041395
NKYFJZAKUPSUSH-UHFFFAOYSA-N
3-bromothioanisole,1-bromo-3-methylthio benzene,3-bromophenyl methyl sulfide,3-bromophenyl methyl sulfane,1-bromo-3-methylsulfanyl benzene,benzene, 1-bromo-3-methylthio,m-bromo methylthio benzene,3-bromophenyl methyl sulphide,1-bromo-3-methylsulfanyl-benzene,3-bromothioanisol
2735627
1-bromo-3-methylsulfanylbenzene
CSC1=CC(=CC=C1)Br
33733-73-2
C7H7BrS
203.097
MFCD00041395
NKYFJZAKUPSUSH-UHFFFAOYSA-N
3-bromothioanisole,1-bromo-3-methylthio benzene,3-bromophenyl methyl sulfide,3-bromophenyl methyl sulfane,1-bromo-3-methylsulfanyl benzene,benzene, 1-bromo-3-methylthio,m-bromo methylthio benzene,3-bromophenyl methyl sulphide,1-bromo-3-methylsulfanyl-benzene,3-bromothioanisol
2735627
1-bromo-3-methylsulfanylbenzene
CSC1=CC(=CC=C1)Br
Boiling Point 124°C to 125°C (10mmHg)
Flash Point >110°C (230°F)
Quantity 5g
Beilstein 2078598
Formula Weight 203.11
Percent Purity 98%
Chemical Name or Material 3-Bromothioanisole

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning