Learn More
3-Bromothiophene, 97%, Thermo Scientific Chemicals
Description
3-Bromothiophene is used in the preparation of derivatives such as thienylenic alpha, μ-diformyl-alpha-oligothiophenes and 3-lithiothiophene. It is also used in the synthesis of 3-lithiothiophene by reacting with n-butyllithium.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
3-Bromothiophene is used in the preparation of derivatives such as thienylenic alpha, µ-diformyl-alpha-oligothiophenes and 3-lithiothiophene. It is also used in the synthesis of 3-lithiothiophene by reacting with n-butyllithium.
Solubility
Immiscible with water.
Notes
Light sensitive. Incompatible with strong oxidizing agents, strong reducing agents and strong bases.
Specifications
Density | 1.722 |
Boiling Point | 157°C to 158°C |
Flash Point | 63°C (145°F) |
Odor | Pungent |
Refractive Index | 1.591 |
Quantity | 10 g |
UN Number | UN2810 |
Beilstein | 105338 |
Solubility Information | Immiscible with water. |
Formula Weight | 163.04 |
Show More |
Safety and Handling
Hazard Category | H226-H301-H310+H330-H315-H319-H335 |
Hazard Statement | GHS H Statement H300-H310-H330-H227-H315-H319-H335 Fatal if swallowed. Fatal in contact with skin. Fatal if inhaled. Combustible liquid. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
Precautionary Statement | P210-P233-P235-P240-P241-P242-P243-P260-P262-P264b-P270-P271-P280-P284-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P363-P370+P378q-P501c |
DOTInformation | Hazard Class: 6.1; Packaging Group: I |
EINECSNumber | 212-821-3 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.