Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Bromothiophene, 97%, ACROS Organics™

Click to view available options
Quantity:
25mL
100mL
500mL
Packaging:
Glass bottle
872-31-1
C4H3BrS
163.03
XCMISAPCWHTVNG-UHFFFAOYSA-N
3-thienyl bromide,thiophene, 3-bromo,3-bromo thiophene,beta-bromothiophene,.beta.-bromothiophene,3-bromo-thiophene,bromothiophene 3-,3-bromothiphene,3-bromthiophene,pubchem5306
13383
3-bromothiophene
C1=CSC=C1Br
872-31-1
C4H3BrS
163.03
XCMISAPCWHTVNG-UHFFFAOYSA-N
3-thienyl bromide,thiophene, 3-bromo,3-bromo thiophene,beta-bromothiophene,.beta.-bromothiophene,3-bromo-thiophene,bromothiophene 3-,3-bromothiphene,3-bromthiophene,pubchem5306
13383
3-bromothiophene
C1=CSC=C1Br
Boiling Point 158.0°C
Density 1.7400g/mL
Infrared Spectrum Authentic
Flash Point 56°C
Refractive Index 1.5905 to 1.5925
Packaging Glass bottle
Specific Gravity 1.74
Quantity 100mL
Color Colorless to Yellow
Assay Percent Range 97%
Solubility Information Solubility in water: immiscible. Other solubilities: soluble in acetone,benzene,slightly soluble in chloroform
Formula Weight 163.03
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material 3-Bromothiophene
Show More Show Less

GHS H Statement
Fatal in contact with skin.
Causes skin irritation.
Fatal if inhaled.
Toxic to aquatic life with long lasting effects.
Causes serious eye irritation.
Toxic if swallowed.
May cause respiratory irritation.
Flammable liquid and vapor.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.
Wash face, hands and any exposed skin thoroughly after handling.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
If skin irritation occurs: Get medical advice/attention.
Wear protective gloves/protective clothing/eye protection/face protection.
If eye irritation persists: Get medical advice/attention.

Recommended Storage : Product may darken on storage