Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-Bromothiophenol, 97%

Click to view available options
Quantity:
1g
5g
25g
6320-01-0
C6H5BrS
189.07
MFCD00009603
HNGQQUDFJDROPY-UHFFFAOYSA-N
3-bromothiophenol,m-bromobenzenethiol,m-bromothiophenol,3-bromo thiophenol,3-bromobenzene-1-thiol,benzenethiol, 3-bromo,3-bromophenyl mercaptan,3-bromo-benzenethiol,pubchem6805,#
80597
3-bromobenzenethiol
C1=CC(=CC(=C1)Br)S
6320-01-0
C6H5BrS
189.07
MFCD00009603
HNGQQUDFJDROPY-UHFFFAOYSA-N
3-bromothiophenol,m-bromobenzenethiol,m-bromothiophenol,3-bromo thiophenol,3-bromobenzene-1-thiol,benzenethiol, 3-bromo,3-bromophenyl mercaptan,3-bromo-benzenethiol,pubchem6805,#
80597
3-bromobenzenethiol
C1=CC(=CC(=C1)Br)S
Flash Point 105°C (221°F)
Odor Unpleasant
Quantity 1g
Boiling Point 119°C to 121°C (21mmHg)
Beilstein 1926242
Formula Weight 189.08
Percent Purity 97%
Chemical Name or Material 3-Bromothiophenol

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning