Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-Butyn-1-ol, 98%

Manufacturer:  Alfa Aesar™ A1147706

Quantity 5g

 View more versions of this product

Catalog No. AAA1147706


  Check Availability
Add to cart

Specifications

Description

Applications
3-butyn-1-ol is used in the study of a palladium-catalyzed coupling with beta-tetrionic acid bromide to prepare alkynyl substituted furanones. It is also used to prepare Markovnikov addition product. Further, it is used as an intermediate of an active pharmaceutical ingredient such as fexofenadine.

Solubility
Miscible with alcohols and organic solvents. Immiscible with aliphatic hydrocarbon solvents.

Notes
Incompatible with strong oxidizing agents, acids, acid anhydrides and copper.
Description & Specifications

Chemical Identifiers

927-74-2
HC≡CCH2CH2OH
MFCD00002955
773710
3-Butyn-1-ol
C4H6O
70.09
UN1987
4-Hydroxy-1-butyne

Specifications

927-74-2
HC≡CCH2CH2OH
MFCD00002955
773710
129° to 130°C
5g
Characteristic
≥98%
C4H6O
70.09
UN1987
4-Hydroxy-1-butyne
−63°C
36°C (96°F)
3-Butyn-1-ol

Safety and Handling

SDS & Safety Documents

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280g-P305+P351+P338
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.

Warning

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALCOHOLS, N.O.S.

RTECSNumber : ES0710000

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit