Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Butynyl p-toluenesulfonate, 96%, Acros Organics

Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass bottle
23418-85-1
C11H12O3S
224.28
MFCD00041687
STOASOOVVADOKH-UHFFFAOYSA-N
3-butynyl p-toluenesulfonate,but-3-yn-1-yl 4-methylbenzenesulfonate,3-butynyl tosylate,3-butynyl 4-toluenesulfonate,3-1-butynyl-4-toluenesulphonate,3-butynyl 4-methylbenzenesulfonate,p-toluenesulfonic acid 3-butynyl ester,but-3-ynyl tosylate,4-tosyloxy-1-butyne,acmc-1cgfq
272020
but-3-ynyl 4-methylbenzenesulfonate
CC1=CC=C(C=C1)S(=O)(=O)OCCC#C
23418-85-1
C11H12O3S
224.28
MFCD00041687
STOASOOVVADOKH-UHFFFAOYSA-N
3-butynyl p-toluenesulfonate,but-3-yn-1-yl 4-methylbenzenesulfonate,3-butynyl tosylate,3-butynyl 4-toluenesulfonate,3-1-butynyl-4-toluenesulphonate,3-butynyl 4-methylbenzenesulfonate,p-toluenesulfonic acid 3-butynyl ester,but-3-ynyl tosylate,4-tosyloxy-1-butyne,acmc-1cgfq
272020
but-3-ynyl 4-methylbenzenesulfonate
CC1=CC=C(C=C1)S(=O)(=O)OCCC#C
Flash Point 110°C
Density 1.0120g/mL
Specific Gravity 1.012
Refractive Index 1.5220 to 1.524
Color Yellow
Infrared Spectrum Authentic
Quantity 1g
Packaging Glass bottle
CAS Min % 95.0
CAS Max % 100.0
Assay Percent Range 95% min. (GC)
Formula Weight 224.28
Physical Form Liquid
Percent Purity 96%
Chemical Name or Material 3-Butynyl p-toluenesulfonate
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning