Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Carbamoyl-2,2,5,5-tetramethylpyrrolidin-1-yloxy, 97%, free radical, ACROS Organics™

Click to view available options
Quantity:
1g
Packaging:
Glass bottle
4399-80-8
C9 H17 N2 O2
MFCD00003168
2,2,5,5-Tetramethyl-1-pyrrolidinyloxy-3-carboxamide
4399-80-8
C9 H17 N2 O2
MFCD00003168
2,2,5,5-Tetramethyl-1-pyrrolidinyloxy-3-carboxamide
Polymerization reagent
Packaging Glass bottle
Melting Point 172°C to 176°C
Infrared Spectrum Authentic
Color Yellow
Quantity 1g
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Formula Weight 185.25
Physical Form Crystalline Solid
Percent Purity ≥96.0%
Chemical Name or Material 3-Carbamoyl-2,2,5,5-tetramethylpyrrolidin-1-yloxy, 97%
Show More Show Less