Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-Carbamoyl-2-fluorobenzeneboronic acid pinacol ester, 96%

Click to view available options
Quantity:
250mg
1g
3-Carbamoyl-2-fluorophenylboronic acid pinacol ester
3-Carbamoyl-2-fluorophenylboronic acid pinacol ester
Quantity 1g
Formula Weight 265.09
Physical Form Solid
Percent Purity 96%
Chemical Name or Material 3-Carbamoyl-2-fluorobenzeneboronic acid pinacol ester

TSCA : No

Recommended Storage : Ambient temperatures