Alfa Aesar™ 3-Carbamoylbenzeneboronic acid, 98%

Manufacturer: Alfa Aesar™ H5286903

Quantity 1g

 View more versions of this product

Catalog No. AAH5286903

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 351422-73-6
    Molecular Formula C7H8BNO3
    Formula Weight 164.96
    MDL Number MFCD03411948
    Synonym 3-Aminocarbonylphenylboronic acid; 3-Carbamoylphenylboronic acid
    Chemical Name or Material 3-Carbamoylbenzeneboronic acid

    Specifications

    Formula Weight 164.96
    Molecular Formula C7H8BNO3
    MDL Number MFCD03411948
    Health Hazard 1 Danger
    Health Hazard 3 GHS P Statement
    P261-P280-P301+P310-P305+P351+P338-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Quantity 1g
    CAS 351422-73-6
    Synonym 3-Aminocarbonylphenylboronic acid; 3-Carbamoylphenylboronic acid
    Health Hazard 2 GHS H Statement
    H301-H315-H319-H335
    Toxic if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Melting Point 232° to 238°C
    Chemical Name or Material 3-Carbamoylbenzeneboronic acid
    Assay 98%

    GHS H Statement
    H301-H315-H319-H335
    Toxic if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P301+P310-P305+P351+P338-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit