Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Carbamoylphenylboronic acid pinacol ester, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
188665-74-9
C13H18BNO3
247.1
BPKIPHYWHVOWMS-UHFFFAOYSA-N
3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzamide,3-aminocarbonylphenylboronic acid, pinacol ester,3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-benzamide,3-aminocarbonylphenylboronic acid pinacol ester,benzamide, 3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl,3-tetramethyl-1,3,2-dioxaborolan-2-yl benzamide,2-dioxaborolan-2-yl-benzamide,bpkiphywhvowms-uhfffaoysa-n,3-carbamoylphenylboronic acid pinacol ester
2760587
188665-74-9
C13H18BNO3
247.1
BPKIPHYWHVOWMS-UHFFFAOYSA-N
3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzamide,3-aminocarbonylphenylboronic acid, pinacol ester,3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-benzamide,3-aminocarbonylphenylboronic acid pinacol ester,benzamide, 3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl,3-tetramethyl-1,3,2-dioxaborolan-2-yl benzamide,2-dioxaborolan-2-yl-benzamide,bpkiphywhvowms-uhfffaoysa-n,3-carbamoylphenylboronic acid pinacol ester
2760587
Melting Point 182°C to 183°C
Packaging Glass Bottle
Quantity 1g
Assay Percent Range 96% min. (GC)
Formula Weight 247.1
Percent Purity 97%
Chemical Name or Material 3-Carbamoylphenylboronic acid pinacol ester

GHS Signal Word: Warning