Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Carboxy-4-fluorophenylboronic acid, 97%, ACROS Organics™

Click to view available options
Quantity:
10g
Packaging:
Glass bottle
872460-12-3
C7H6BFO4
183.93
MFCD03095129
DGORTXQWDDXSIQ-UHFFFAOYSA-N
3-carboxy-4-fluorophenylboronic acid,3-carboxy-4-fluorobenzeneboronic acid,5-dihydroxyboranyl-2-fluorobenzoic acid,4-fluoro-3-carboxyphenylboronic acid,benzoic acid, 5-borono-2-fluoro,3-carboxyl-4-fluorophenylboronic acid,5-borono-2-fluoro-benzoic acid,akos brn-0540,pubchem16787,acmc-209qj6
2763242
5-borono-2-fluorobenzoic acid
B(C1=CC(=C(C=C1)F)C(=O)O)(O)O
872460-12-3
C7H6BFO4
183.93
MFCD03095129
DGORTXQWDDXSIQ-UHFFFAOYSA-N
3-carboxy-4-fluorophenylboronic acid,3-carboxy-4-fluorobenzeneboronic acid,5-dihydroxyboranyl-2-fluorobenzoic acid,4-fluoro-3-carboxyphenylboronic acid,benzoic acid, 5-borono-2-fluoro,3-carboxyl-4-fluorophenylboronic acid,5-borono-2-fluoro-benzoic acid,akos brn-0540,pubchem16787,acmc-209qj6
2763242
5-borono-2-fluorobenzoic acid
B(C1=CC(=C(C=C1)F)C(=O)O)(O)O
Quantity 10g
Packaging Glass bottle
Melting Point 196.0°C to 204.0°C
Color White
Infrared Spectrum Authentic
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (HPLC)
Formula Weight 183.93
Physical Form Crystalline Solid
Percent Purity 97%
Chemical Name or Material 3-Carboxy-4-fluorophenylboronic acid
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Harmful if swallowed.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning