Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-Carboxybenzeneboronic acid

Click to view available options
Quantity:
1g
5g
25487-66-5
C7H7BO4
165.939
MFCD00036833
DBVFWZMQJQMJCB-UHFFFAOYSA-N
3-carboxyphenylboronic acid,3-dihydroxyboranyl benzoic acid,3-carboxybenzeneboronic acid,m-carboxyphenylboronic acid,3-dihydroxyborane benzoic acid,benzoic acid, 3-borono,3-dihydroxyboryl benzoic acid,3-phenyl ester boronic acid,3-carboxyphenyl boronic acid,3-carboxy-phenyl-boronic acid
2733957
3-boronobenzoic acid
B(C1=CC(=CC=C1)C(=O)O)(O)O
25487-66-5
C7H7BO4
165.939
MFCD00036833
DBVFWZMQJQMJCB-UHFFFAOYSA-N
3-carboxyphenylboronic acid,3-dihydroxyboranyl benzoic acid,3-carboxybenzeneboronic acid,m-carboxyphenylboronic acid,3-dihydroxyborane benzoic acid,benzoic acid, 3-borono,3-dihydroxyboryl benzoic acid,3-phenyl ester boronic acid,3-carboxyphenyl boronic acid,3-carboxy-phenyl-boronic acid
2733957
3-boronobenzoic acid
B(C1=CC(=CC=C1)C(=O)O)(O)O
Melting Point ~240°C (decomposition)
Quantity 1g
Beilstein 2967400
Formula Weight 165.94
Chemical Name or Material 3-Carboxybenzeneboronic acid

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : CY8750000