Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
3-(Carboxymethyl)benzothiazolium Bromide 97.0+%, TCI America™
Click to view available options
Quantity:
25g
Specifications
Color | Yellow |
Quantity | 25g |
Formula Weight | 274.13 |
Physical Form | Crystal-Powder at 20°C |
Percent Purity | ≥97.0% (T) |
Chemical Name or Material | 3-(Carboxymethyl)benzothiazolium Bromide |
Safety and Handling
TSCA : No