Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Carboxyphenylboronic acid, 97%, Acros Organics™

Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass bottle
25487-66-5
C7H7BO4
165.94
MFCD00036833
DBVFWZMQJQMJCB-UHFFFAOYSA-N
3-carboxyphenylboronic acid,3-dihydroxyboranyl benzoic acid,3-carboxybenzeneboronic acid,m-carboxyphenylboronic acid,3-dihydroxyborane benzoic acid,benzoic acid, 3-borono,3-dihydroxyboryl benzoic acid,3-phenyl ester boronic acid,3-carboxyphenyl boronic acid,3-carboxy-phenyl-boronic acid
2733957
3-boronobenzoic acid
B(C1=CC(=CC=C1)C(=O)O)(O)O
25487-66-5
C7H7BO4
165.94
MFCD00036833
DBVFWZMQJQMJCB-UHFFFAOYSA-N
3-carboxyphenylboronic acid,3-dihydroxyboranyl benzoic acid,3-carboxybenzeneboronic acid,m-carboxyphenylboronic acid,3-dihydroxyborane benzoic acid,benzoic acid, 3-borono,3-dihydroxyboryl benzoic acid,3-phenyl ester boronic acid,3-carboxyphenyl boronic acid,3-carboxy-phenyl-boronic acid
2733957
3-boronobenzoic acid
B(C1=CC(=CC=C1)C(=O)O)(O)O
Packaging Glass bottle
Melting Point 243.0°C to 247.0°C
Quantity 5g
Color White
Infrared Spectrum Authentic
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97%
Linear Formula HOOCC6H4B(OH)2
Solubility Information Solubility in water: +/- 2.5%.
Formula Weight 165.94
Physical Form Crystalline Powder
Percent Purity 97%
Chemical Name or Material 3-Carboxyphenylboronic acid
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.