Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
(±)-3-Chloro-1,2-propanediol, Honeywell Fluka™
Click to view available options
Quantity:
100 mg
Description
Ensure you have the information you need to quickly start your analysis, and pass relevant audits:
- High quality inorganic and organic reference materials produced according to ISO 9001
- Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results
- Free expert technical support
Specifications
Physical Form | Viscous Liquid |
Grade | Analytical Standard |
Packaging | Glass Bottle |
Density | 1.32 g/cm3 |
Boiling Point | 216°C to 219°C |
Quantity | 100 mg |
Flash Point | 113°C |
For Use With (Application) | ALIPHATICS |
Safety and Handling
P260+P280+P201
H372+H370+H360+H351+H330+H301+H315+H318
Danger