Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Chloro-1,2-propanediol, 99%, Thermo Scientific™

Click to view available options
Quantity:
25mL
500mL
1L
2.5L
Packaging:
Glass bottle
96-24-2
C3H7ClO2
110.54
MFCD00004712
SSZWWUDQMAHNAQ-UHFFFAOYNA-N
3-chloro-1,2-propanediol, alpha-chlorohydrin, chlorodeoxyglycerol, glycerol alpha-monochlorohydrin, epibloc, 1,2-propanediol, 3-chloro, a-chlorohydrin, 3-chloropropanediol, 3-chloropropylene glycol, glycerol chlorohydrin
7290
CHEBI:18721
3-chloropropane-1,2-diol
OCC(O)CCl
96-24-2
C3H7ClO2
110.54
MFCD00004712
SSZWWUDQMAHNAQ-UHFFFAOYNA-N
3-chloro-1,2-propanediol, alpha-chlorohydrin, chlorodeoxyglycerol, glycerol alpha-monochlorohydrin, epibloc, 1,2-propanediol, 3-chloro, a-chlorohydrin, 3-chloropropanediol, 3-chloropropylene glycol, glycerol chlorohydrin
7290
CHEBI:18721
3-chloropropane-1,2-diol
OCC(O)CCl

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

CAS Min % 98.5
CAS Max % 100.0
Density 1.3220g/mL
Color Yellow
Melting Point -40.0°C
Boiling Point 213.0°C
Flash Point 135°C
Assay Percent Range 98.5% min. (GC)
Infrared Spectrum Authentic
Linear Formula ClCH2CH(OH)CH2OH
Packaging Glass bottle
Merck Index 15, 2146
Refractive Index 1.4790 to 1.4810
Quantity 2.5L
Solubility Information Solubility in water: soluble. Other solubilities: soluble in ethanol,acetone,methanol and ether,slightly soluble in toluene
Specific Gravity 1.322
Formula Weight 110.54
Physical Form Liquid
Percent Purity 99%
Viscosity 215 mPa.s (20°C)
Chemical Name or Material 3-Chloro-1, 2-propanediol
Show More Show Less

GHS H Statement
Suspected of causing genetic defects.
Fatal if swallowed.
Toxic in contact with skin.
Causes serious eye irritation.
Fatal if inhaled.
May cause respiratory irritation.
Causes skin irritation.<br

GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN:

GHS Signal Word: Danger

EINECSNumber : 202-492-4

RUO – Research Use Only