Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-chloro-2,4,5-trifluorobenzoic acid, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
Packaging:
Glass bottle
101513-77-3
C7H2ClF3O2
210.536
MFCD00153101
KBESHLYCSZINAJ-UHFFFAOYSA-N
2,4,5-trifluoro-3-chlorobenzoic acid, 3-chloro-2,4,5-trifluoro-benzoic acid, 3-chloro-2,4,5-trifluorobenzoicacid, benzoic acid, 3-chloro-2,4,5-trifluoro, pubchem1376, acmc-1bvtw, ksc502q6n, 3-chloro-2,4,5trifluorobenzoic acid, 3-chloro-2,4,5-trilfuorobenzoic acid, 3-carboxy-2,5,6-trifluorochlorobenzene
2734269
3-chloro-2,4,5-trifluorobenzoic acid
C1=C(C(=C(C(=C1F)F)Cl)F)C(=O)O
101513-77-3
C7H2ClF3O2
210.536
MFCD00153101
KBESHLYCSZINAJ-UHFFFAOYSA-N
2,4,5-trifluoro-3-chlorobenzoic acid, 3-chloro-2,4,5-trifluoro-benzoic acid, 3-chloro-2,4,5-trifluorobenzoicacid, benzoic acid, 3-chloro-2,4,5-trifluoro, pubchem1376, acmc-1bvtw, ksc502q6n, 3-chloro-2,4,5trifluorobenzoic acid, 3-chloro-2,4,5-trilfuorobenzoic acid, 3-carboxy-2,5,6-trifluorochlorobenzene
2734269
3-chloro-2,4,5-trifluorobenzoic acid
C1=C(C(=C(C(=C1F)F)Cl)F)C(=O)O
Packaging Glass bottle
Color White
Infrared Spectrum Authentic
Quantity 5g
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula F3C6H(Cl)CO2H
Formula Weight 210.54
Physical Form Powder
Percent Purity ≥97.5%
Chemical Name or Material 3-Chloro-2, 4, 5-trifluorobenzoic acid, 98%
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.