Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-Chloro-2,4-difluorobenzoic acid, 97%

Click to view available options
Quantity:
1g
5g
25g
154257-75-7
C7H3ClF2O2
192.546
MFCD01631387
YGYZTZAEZMCPSC-UHFFFAOYSA-N
3-chloro-2,4-difluoro-benzoic acid,2,4-difluoro-3-chlorobenzoic acid,3-chloro-2,4-difluorobenzoicacid,benzoic acid, 3-chloro-2,4-difluoro,pubchem2217,acmc-1byme,intermediates-zcf02173,ksc181k3p
2773532
3-chloro-2,4-difluorobenzoic acid
C1=CC(=C(C(=C1C(=O)O)F)Cl)F
154257-75-7
C7H3ClF2O2
192.546
MFCD01631387
YGYZTZAEZMCPSC-UHFFFAOYSA-N
3-chloro-2,4-difluoro-benzoic acid,2,4-difluoro-3-chlorobenzoic acid,3-chloro-2,4-difluorobenzoicacid,benzoic acid, 3-chloro-2,4-difluoro,pubchem2217,acmc-1byme,intermediates-zcf02173,ksc181k3p
2773532
3-chloro-2,4-difluorobenzoic acid
C1=CC(=C(C(=C1C(=O)O)F)Cl)F
Melting Point 174°C to 177°C
Quantity 1g
Beilstein 7290655
Formula Weight 192.55
Percent Purity 97%
Chemical Name or Material 3-Chloro-2,4-difluorobenzoic acid

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 000-000-0

TSCA : No

Recommended Storage : Ambient temperatures