Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Chloro-2,4-pentanedione, 98%, ACROS Organics™

Click to view available options
Quantity:
5mL
100mL
500mL
2.5L
1694-29-7
C5H7ClO2
134.56
MFCD00009651
VLRGXXKFHVJQOL-UHFFFAOYSA-N
3-chloro-2,4-pentanedione,3-chloroacetylacetone,2,4-pentanedione, 3-chloro,1-acetyl-1-chloroacetone,3-chloro 2,4-pentanedione,3-chloro.2,4-pentanedione,3-chloro-pentane-2,4-dione,2,4-pentanedione,3-chloro,3-chloro-2,4-pentanedione, produced by wacker chemie ag, burghausen, germany gc
74328
3-chloropentane-2,4-dione
CC(=O)C(C(=O)C)Cl
1694-29-7
C5H7ClO2
134.56
MFCD00009651
VLRGXXKFHVJQOL-UHFFFAOYSA-N
3-chloro-2,4-pentanedione,3-chloroacetylacetone,2,4-pentanedione, 3-chloro,1-acetyl-1-chloroacetone,3-chloro 2,4-pentanedione,3-chloro.2,4-pentanedione,3-chloro-pentane-2,4-dione,2,4-pentanedione,3-chloro,3-chloro-2,4-pentanedione, produced by wacker chemie ag, burghausen, germany gc
74328
3-chloropentane-2,4-dione
CC(=O)C(C(=O)C)Cl
Melting Point -15°C
Boiling Point 49°C to 52°C (18.0mmHg)
Specific Gravity 1.179
Flash Point 56°C
Refractive Index 1.4825 to 1.4845
Density 1.1790g/mL
Color Brown to Yellow
Quantity 2.5L
Infrared Spectrum Authentic
Packaging Glass bottle
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (GC)
Linear Formula CH3COCH(Cl)COCH3
Beilstein 01, 785
Solubility Information Solubility in water: 7.65g/L (20°C). Other solubilities: soluble in organic solvents
Formula Weight 134.56
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material 3-Chloro-2, 4-pentanedione, 98%
Show More Show Less

GHS H Statement
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.
Harmful in contact with skin.
May cause respiratory irritation.
Flammable liquid and vapor.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor

GHS Signal Word: Warning

EINECSNumber : 216-902-4

TSCA : TSCA