Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3'-Chloro-2',6'-difluoroacetophenone, 97+%

Click to view available options
Quantity:
1g
5g
177942-50-6
C8H5ClF2O
190.574
MFCD01631329
GZGJXQFBPXHWRR-UHFFFAOYSA-N
3'-chloro-2',6'-difluoroacetophenone,1-3-chloro-2,6-difluorophenyl ethanone,2',6'-difluoro-3'-chloroacetophenone,3-chloro-2,6-difluoroacetophenone,1-3-chloro-2,6-difluorophenyl ethan-1-one,1-3-chloro-2,6-difluoro-phenyl-ethanone,1-acetyl-3-chloro-2,6-difluorobenzene,2,6-difluoro-3-chloroacetophenone,pubchem4317,pubchem7092
2773505
1-(3-chloro-2,6-difluorophenyl)ethanone
CC(=O)C1=C(C=CC(=C1F)Cl)F
177942-50-6
C8H5ClF2O
190.574
MFCD01631329
GZGJXQFBPXHWRR-UHFFFAOYSA-N
3'-chloro-2',6'-difluoroacetophenone,1-3-chloro-2,6-difluorophenyl ethanone,2',6'-difluoro-3'-chloroacetophenone,3-chloro-2,6-difluoroacetophenone,1-3-chloro-2,6-difluorophenyl ethan-1-one,1-3-chloro-2,6-difluoro-phenyl-ethanone,1-acetyl-3-chloro-2,6-difluorobenzene,2,6-difluoro-3-chloroacetophenone,pubchem4317,pubchem7092
2773505
1-(3-chloro-2,6-difluorophenyl)ethanone
CC(=O)C1=C(C=CC(=C1F)Cl)F
Applications
It is an intermediate in organic syntheses.

Notes
Store at room temperature. Incompatible with oxidizing agents.
Quantity 5g
Formula Weight 190.58
Percent Purity ≥97%
Chemical Name or Material 3'-Chloro-2',6'-difluoroacetophenone

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures