Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-Chloro-2,6-dimethylaniline, 99%

Click to view available options
Quantity:
10g
50g
250g
26829-77-6
C8H10ClN
155.625
MFCD02683086
FGMXFOTYCHZCLA-UHFFFAOYSA-N
3-chloro-2,6-dimethyl-phenylamine,acmc-20an7h,3-chloro-2,6-xylidine,3-chloro-2,6-dimethylphenylamine,3-chloranyl-2,6-dimethyl-aniline,benzenamine,3-chloro-2,6-dimethyl
7023016
3-chloro-2,6-dimethylaniline
CC1=C(C(=C(C=C1)Cl)C)N
26829-77-6
C8H10ClN
155.625
MFCD02683086
FGMXFOTYCHZCLA-UHFFFAOYSA-N
3-chloro-2,6-dimethyl-phenylamine,acmc-20an7h,3-chloro-2,6-xylidine,3-chloro-2,6-dimethylphenylamine,3-chloranyl-2,6-dimethyl-aniline,benzenamine,3-chloro-2,6-dimethyl
7023016
3-chloro-2,6-dimethylaniline
CC1=C(C(=C(C=C1)Cl)C)N
Applications
3-cyano- 2,4-dimethylchlorobenzene was obtained from 3-Chloro-2,6-dimethylaniline which upon hydrolysis gives acid.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Quantity 50g
Melting Point 30°C to 32°C
Boiling Point 136°C to 138°C (17mmHg)
Formula Weight 155.63
Percent Purity 99%
Chemical Name or Material 3-Chloro-2,6-dimethylaniline

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger