Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-Chloro-2,6-dimethylaniline hydrochloride, 99%

Click to view available options
Quantity:
5g
25g
72725-98-5
C8H11Cl2N
192.083
MFCD02683075
WNBACZJGRRMILU-UHFFFAOYSA-N
3-chloro-2,6-dimethylaniline hydrochloride,3-chloro-2,6-dimethylaniline hcl,3-chloro-2,6-xylidine hydrochloride,3-chloro-2,6-dimethyl aniline hydrochloride,3-chloro-2,6-dimethylaniline-hydrogen chloride 1/1
12448951
3-chloro-2,6-dimethylaniline;hydrochloride
CC1=C(C(=C(C=C1)Cl)C)N.Cl
72725-98-5
C8H11Cl2N
192.083
MFCD02683075
WNBACZJGRRMILU-UHFFFAOYSA-N
3-chloro-2,6-dimethylaniline hydrochloride,3-chloro-2,6-dimethylaniline hcl,3-chloro-2,6-xylidine hydrochloride,3-chloro-2,6-dimethyl aniline hydrochloride,3-chloro-2,6-dimethylaniline-hydrogen chloride 1/1
12448951
3-chloro-2,6-dimethylaniline;hydrochloride
CC1=C(C(=C(C=C1)Cl)C)N.Cl
Melting Point ~240°C (decomposition)
Quantity 25g
Formula Weight 192.09
Percent Purity 99%
Chemical Name or Material 3-Chloro-2,6-dimethylaniline hydrochloride

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger