Alfa Aesar™ 3-Chloro-2-fluoro-5-(trifluoromethyl)aniline, 97%

Manufacturer: Alfa Aesar™ H3228103

Quantity 1g

 View more versions of this product

Catalog No. AAH3228103

  Check Availability
Add to Cart

    Chemical Identifiers

    Molecular Formula C7H4ClF4N
    Formula Weight 213.56
    MDL Number MFCD11519322
    UN Number UN2810
    Synonym 3-Amino-5-chloro-2-fluorobenzotrifluoride
    Chemical Name or Material 3-Chloro-2-fluoro-5-(trifluoromethyl)aniline

    Specifications

    Formula Weight 213.56
    UN Number UN2810
    Synonym 3-Amino-5-chloro-2-fluorobenzotrifluoride
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H311-H302-H332-H315-H319
    Toxic in contact with skin.
    Harmful if swallowed.
    Harmful if inhaled.
    Causes skin irritation.
    Causes serious eye irritation.
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
    Quantity 1g
    Molecular Formula C7H4ClF4N
    MDL Number MFCD11519322
    Health Hazard 3 GHS P Statement
    P280h-P305+P351+P338-P309-P310-P302+P352
    Wear protective gloves/clothing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN: Wash with plenty of soap and water.
    Chemical Name or Material 3-Chloro-2-fluoro-5-(trifluoromethyl)aniline
    Assay 97%

    GHS H Statement
    H311-H302-H332-H315-H319
    Toxic in contact with skin.
    Harmful if swallowed.
    Harmful if inhaled.
    Causes skin irritation.
    Causes serious eye irritation.

    GHS P Statement
    P280h-P305+P351+P338-P309-P310-P302+P352
    Wear protective gloves/clothing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN: Wash with plenty of soap and water.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit