Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-Chloro-2-fluoro-5-(trifluoromethyl)benzaldehyde, 97%

Click to view available options
Quantity:
1g
5g
261763-02-4
C8H3ClF4O
226.555
MFCD01631510
MSZTVIFIFJCNRQ-UHFFFAOYSA-N
3-chloro-2-fluoro-5-trifluoromethyl benzaldehyde,benzaldehyde, 3-chloro-2-fluoro-5-trifluoromethyl,3-chloro-2-fluoro-5-trifluoromethyl-benzaldehyde,pubchem2887,acmc-20an7f,3-chloro-5-trifluoromethyl-2-fluorobenzaldehyde,3-chloranyl-2-fluoranyl-5-trifluoromethyl benzaldehyde
736336
3-chloro-2-fluoro-5-(trifluoromethyl)benzaldehyde
C1=C(C=C(C(=C1Cl)F)C=O)C(F)(F)F
261763-02-4
C8H3ClF4O
226.555
MFCD01631510
MSZTVIFIFJCNRQ-UHFFFAOYSA-N
3-chloro-2-fluoro-5-trifluoromethyl benzaldehyde,benzaldehyde, 3-chloro-2-fluoro-5-trifluoromethyl,3-chloro-2-fluoro-5-trifluoromethyl-benzaldehyde,pubchem2887,acmc-20an7f,3-chloro-5-trifluoromethyl-2-fluorobenzaldehyde,3-chloranyl-2-fluoranyl-5-trifluoromethyl benzaldehyde
736336
3-chloro-2-fluoro-5-(trifluoromethyl)benzaldehyde
C1=C(C=C(C(=C1Cl)F)C=O)C(F)(F)F
Flash Point 96°C (204°F)
Quantity 1g
Boiling Point 196°C
Formula Weight 226.56
Percent Purity 97%
Chemical Name or Material 3-Chloro-2-fluoro-5-(trifluoromethyl)benzaldehyde

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning