Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-Chloro-2-fluoro-5-(trifluoromethyl)benzoic acid, 97%

Click to view available options
Quantity:
1g
5g
129931-45-9
C8H3ClF4O2
242.554
MFCD01631509
UINXNXRHKMRASA-UHFFFAOYSA-N
3-chloro-2-fluoro-5-trifluoromethyl benzoic acid,rarechem al be 0499,3-chloro-2-fluoro-5-trifluoromethyl-benzoic acid,pubchem4559,acmc-20a41y,2-fluoro-3-chloro-5-trifluoromethyl-benzoic acid,benzoic acid,3-chloro-2-fluoro-5-trifluoromethyl,3-chloro-2-fluoro-5-trifluoromethyl benzoicacid
2773751
3-chloro-2-fluoro-5-(trifluoromethyl)benzoic acid
C1=C(C=C(C(=C1Cl)F)C(=O)O)C(F)(F)F
129931-45-9
C8H3ClF4O2
242.554
MFCD01631509
UINXNXRHKMRASA-UHFFFAOYSA-N
3-chloro-2-fluoro-5-trifluoromethyl benzoic acid,rarechem al be 0499,3-chloro-2-fluoro-5-trifluoromethyl-benzoic acid,pubchem4559,acmc-20a41y,2-fluoro-3-chloro-5-trifluoromethyl-benzoic acid,benzoic acid,3-chloro-2-fluoro-5-trifluoromethyl,3-chloro-2-fluoro-5-trifluoromethyl benzoicacid
2773751
3-chloro-2-fluoro-5-(trifluoromethyl)benzoic acid
C1=C(C=C(C(=C1Cl)F)C(=O)O)C(F)(F)F
Applications
3-Chloro-2-fluoro-5-(trifluoromethyl)benzoic acid is used as a pharmaceutical intermediate.

Solubility
Soluble in most organic solvents, basic aqueous solutions.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible material is strong oxidizing agents.
Melting Point 89°C to 91°C
Quantity 1g
Formula Weight 242.56
Percent Purity 97%
Chemical Name or Material 3-Chloro-2-fluoro-5-(trifluoromethyl)benzoic acid

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning