Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-Chloro-2-fluoro-5-(trifluoromethyl)phenol, 97+%

Click to view available options
Quantity:
1g
5g
261763-12-6
C7H3ClF4O
214.544
MFCD01631514
OKLHLYVTNGBKCI-UHFFFAOYSA-N
3-chloro-2-fluoro-5-trifluoromethyl phenol,acmc-20an7d,phenol,3-chloro-2-fluoro-5-trifluoromethyl
2773771
3-chloro-2-fluoro-5-(trifluoromethyl)phenol
C1=C(C=C(C(=C1O)F)Cl)C(F)(F)F
261763-12-6
C7H3ClF4O
214.544
MFCD01631514
OKLHLYVTNGBKCI-UHFFFAOYSA-N
3-chloro-2-fluoro-5-trifluoromethyl phenol,acmc-20an7d,phenol,3-chloro-2-fluoro-5-trifluoromethyl
2773771
3-chloro-2-fluoro-5-(trifluoromethyl)phenol
C1=C(C=C(C(=C1O)F)Cl)C(F)(F)F
Quantity 5g
UN Number UN3265
Formula Weight 214.55
Percent Purity ≥97%
Chemical Name or Material 3-Chloro-2-fluoro-5-(trifluoromethyl)phenol

GHS H Statement
H314-H318-H302-H312
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.

GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.