Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-Chloro-2-fluorobenzaldehyde, 97%

Click to view available options
Quantity:
1g
5g
25g
85070-48-0
C7H4ClFO
158.556
MFCD01631571
YAOZCMANASAVFN-UHFFFAOYSA-N
2-fluoro-3-chlorobenzaldehyde,2-fluoro-3-chloro-benzaldehyde,3-chloro-2-fluoro-benzaldehyde,benzaldehyde, 3-chloro-2-fluoro,pubchem1408,acmc-209q3u,2-fluoro chloro benzaldehyde,ksc496e9b,3-chloro-2-fluoro-benzoaldehyde,timtec-bb sbb003987
736335
3-chloro-2-fluorobenzaldehyde
C1=CC(=C(C(=C1)Cl)F)C=O
85070-48-0
C7H4ClFO
158.556
MFCD01631571
YAOZCMANASAVFN-UHFFFAOYSA-N
2-fluoro-3-chlorobenzaldehyde,2-fluoro-3-chloro-benzaldehyde,3-chloro-2-fluoro-benzaldehyde,benzaldehyde, 3-chloro-2-fluoro,pubchem1408,acmc-209q3u,2-fluoro chloro benzaldehyde,ksc496e9b,3-chloro-2-fluoro-benzoaldehyde,timtec-bb sbb003987
736335
3-chloro-2-fluorobenzaldehyde
C1=CC(=C(C(=C1)Cl)F)C=O
Quantity 5g
Beilstein 5861249
Formula Weight 158.56
Percent Purity 97%
Chemical Name or Material 3-Chloro-2-fluorobenzaldehyde

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning