Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-Chloro-2-hydroxy-5-(trifluoromethyl)pyridine, 95%

Click to view available options
Quantity:
1g
5g
76041-71-9
C6H3ClF3NO
197.541
MFCD00153095
AJPOOWWMZOPUCG-UHFFFAOYSA-N
3-chloro-5-trifluoromethyl pyridin-2-ol,3-chloro-2-hydroxy-5-trifluoromethyl pyridine,3-chloro-5-trifluoromethyl-2-pyridinol,3-chloro-5-trifluoromethyl-1,2-dihydropyridin-2-one,3-chloro-5-trifluoromethyl-1h-pyridin-2-one,2 1h-pyridinone, 3-chloro-5-trifluoromethyl,2-hydroxy-3-chloro-5-trifluoromethyl pyridine,3-chloro-5-trifluoromethyl pyridin-2 1h-one,3-chloro-5-trifluoromethyl hydropyridin-2-one,pubchem23825
725436
3-chloro-5-(trifluoromethyl)-1H-pyridin-2-one
C1=C(C(=O)NC=C1C(F)(F)F)Cl
76041-71-9
C6H3ClF3NO
197.541
MFCD00153095
AJPOOWWMZOPUCG-UHFFFAOYSA-N
3-chloro-5-trifluoromethyl pyridin-2-ol,3-chloro-2-hydroxy-5-trifluoromethyl pyridine,3-chloro-5-trifluoromethyl-2-pyridinol,3-chloro-5-trifluoromethyl-1,2-dihydropyridin-2-one,3-chloro-5-trifluoromethyl-1h-pyridin-2-one,2 1h-pyridinone, 3-chloro-5-trifluoromethyl,2-hydroxy-3-chloro-5-trifluoromethyl pyridine,3-chloro-5-trifluoromethyl pyridin-2 1h-one,3-chloro-5-trifluoromethyl hydropyridin-2-one,pubchem23825
725436
3-chloro-5-(trifluoromethyl)-1H-pyridin-2-one
C1=C(C(=O)NC=C1C(F)(F)F)Cl
Melting Point 159°C to 161°C
Quantity 5g
Beilstein 5932693
Formula Weight 197.54
Physical Form Solid
Percent Purity 95%
Chemical Name or Material 3-Chloro-2-hydroxy-5-(trifluoromethyl)pyridine