Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3′-Chloro-2′-methylacetanilide, 98%, Alfa Aesar™

Click to view available options
Quantity:
25g
100g
7463-35-6
C9H10ClNO
183.635
MFCD00018264
NALGTKRTIJHBBK-UHFFFAOYSA-N
n-3-chloro-2-methylphenyl acetamide,3-chloro-2-methylacetanilide,acetamide, n-3-chloro-2-methylphenyl,3'-chloro-2'-methylacetanilide,3-chloro-2-methyl-n-acetylaniline,3'-chloro-o-acetotoluidide,3'-chloro-ortho-acetotoluidide,n-acetyl-3-chloro-2-methylaniline,2-acetamido-6-chlorotoluene,n-2-methyl-3-chlorophenyl acetamide
81969
N-(3-chloro-2-methylphenyl)acetamide
CC1=C(C=CC=C1Cl)NC(=O)C
7463-35-6
C9H10ClNO
183.635
MFCD00018264
NALGTKRTIJHBBK-UHFFFAOYSA-N
n-3-chloro-2-methylphenyl acetamide,3-chloro-2-methylacetanilide,acetamide, n-3-chloro-2-methylphenyl,3'-chloro-2'-methylacetanilide,3-chloro-2-methyl-n-acetylaniline,3'-chloro-o-acetotoluidide,3'-chloro-ortho-acetotoluidide,n-acetyl-3-chloro-2-methylaniline,2-acetamido-6-chlorotoluene,n-2-methyl-3-chlorophenyl acetamide
81969
N-(3-chloro-2-methylphenyl)acetamide
CC1=C(C=CC=C1Cl)NC(=O)C
Packaging Plastic bottle
Melting Point 155°C to 157°C
Quantity 100g
Formula Weight 183.64
Percent Purity 98%
Chemical Name or Material 3′-Chloro-2′-methylacetanilide