Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
3′-Chloro-2′-methylacetanilide, 98%, Alfa Aesar™
Click to view available options
Quantity:
25g
100g
Specifications
Packaging | Plastic bottle |
Quantity | 25g |
Melting Point | 155°C to 157°C |
Formula Weight | 183.64 |
Percent Purity | 98% |
Chemical Name or Material | 3′-Chloro-2′-methylacetanilide |
Safety and Handling
EINECSNumber : 231-254-2
TSCA : No
Recommended Storage : Ambient temperatures