Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Chloro-2-methylaniline 99%, ACROS Organics™

Click to view available options
Quantity:
100mL
500mL
Packaging:
Glass bottle
87-60-5
C7H8ClN
141.598
MFCD00007766
ZUVPLKVDZNDZCM-UHFFFAOYSA-N
3-chloro-o-toluidine,2-amino-6-chlorotoluene,benzenamine, 3-chloro-2-methyl,3-chloro-2-toluidine,scarlet tr base,fast scarlet tr base,6-chloro-2-aminotoluene,o-toluidine, 3-chloro,3-chloro-2-methylphenylamine,o-methyl-m-chloroaniline
6894
3-chloro-2-methylaniline
CC1=C(C=CC=C1Cl)N
87-60-5
C7H8ClN
141.598
MFCD00007766
ZUVPLKVDZNDZCM-UHFFFAOYSA-N
3-chloro-o-toluidine,2-amino-6-chlorotoluene,benzenamine, 3-chloro-2-methyl,3-chloro-2-toluidine,scarlet tr base,fast scarlet tr base,6-chloro-2-aminotoluene,o-toluidine, 3-chloro,3-chloro-2-methylphenylamine,o-methyl-m-chloroaniline
6894
3-chloro-2-methylaniline
CC1=C(C=CC=C1Cl)N
Packaging Glass bottle
Quantity 100mL
Refractive Index 1.5870 to 1.5890
Infrared Spectrum Authentic
Color Brown-Red or Red to Yellow
Melting Point 2.0°C
Flash Point >110°C
Density 1.1710g/mL
Boiling Point 245.0°C
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Linear Formula ClC6H3(CH3)NH2
Beilstein 12, 836
Formula Weight 141.6
Physical Form Liquid
Percent Purity 99%
Chemical Name or Material 3-Chloro-2-methylaniline, 99%
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
Harmful if swallowed.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 201-756-6

RTECSNumber : XU4760000

TSCA : TSCA

Recommended Storage : Product may darken during storage