Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-chloro-2-methylbenzenesulfonyl chloride, 98%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
80563-86-6
C7H6Cl2O2S
225.09
MFCD00052198
ZSIYKAQPQRTBPF-UHFFFAOYSA-N
3-chloro-2-methylbenzene-1-sulfonyl chloride, 3-chloro-2-methylbenzenesulphonyl chloride, benzenesulfonyl chloride, 3-chloro-2-methyl-9ci, 3-chloro-2-methylphenylsulfonyl chloride, 3-chloro-2-methyl-benzenesulfonyl chloride, chloro 3-chloro-2-methylphenyl sulfone, pubchem5096, acmc-209pjq, buttpark 3711-61, 3-chloro-2-methylbenzenesulfonylchloride
522734
3-chloro-2-methylbenzenesulfonyl chloride
CC1=C(C=CC=C1Cl)S(=O)(=O)Cl
80563-86-6
C7H6Cl2O2S
225.09
MFCD00052198
ZSIYKAQPQRTBPF-UHFFFAOYSA-N
3-chloro-2-methylbenzene-1-sulfonyl chloride, 3-chloro-2-methylbenzenesulphonyl chloride, benzenesulfonyl chloride, 3-chloro-2-methyl-9ci, 3-chloro-2-methylphenylsulfonyl chloride, 3-chloro-2-methyl-benzenesulfonyl chloride, chloro 3-chloro-2-methylphenyl sulfone, pubchem5096, acmc-209pjq, buttpark 3711-61, 3-chloro-2-methylbenzenesulfonylchloride
522734
3-chloro-2-methylbenzenesulfonyl chloride
CC1=C(C=CC=C1Cl)S(=O)(=O)Cl
Quantity 5g
Melting Point 71°C to 73°C
Packaging Glass bottle
Infrared Spectrum Authentic
Color White to Yellow
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (ex Cl) (Argentometry)
Linear Formula Cl(CH3)C6H3SO2Cl
Formula Weight 225.09
Physical Form Crystals
Percent Purity 98%
Chemical Name or Material 3-chloro-2-methylbenzenesulfonyl chloride
Show More Show Less

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger