Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Chloro-2-methylbenzoic acid, 98%, Acros Organics™

Click to view available options
Quantity:
1g
5g
7499-08-3
C8H7ClO2
170.6
HXGHMCLCSPQMOR-UHFFFAOYSA-N
3-chloro-o-toluic acid,3-chloro-2-methyl-benzoic acid,benzoic acid, 3-chloro-2-methyl,o-toluic acid, 3-chloro,3-chloro-2-methylbenzoicacid,2-methyl-3-chlorobenzoic acid,pubchem8362,2-carboxy-6-chlorotoluene,3chloro-2-methylbenzoic acid,3-chloro-2-methybenzoic acid
82010
3-chloro-2-methylbenzoic acid
CC1=C(C=CC=C1Cl)C(=O)O
7499-08-3
C8H7ClO2
170.6
HXGHMCLCSPQMOR-UHFFFAOYSA-N
3-chloro-o-toluic acid,3-chloro-2-methyl-benzoic acid,benzoic acid, 3-chloro-2-methyl,o-toluic acid, 3-chloro,3-chloro-2-methylbenzoicacid,2-methyl-3-chlorobenzoic acid,pubchem8362,2-carboxy-6-chlorotoluene,3chloro-2-methylbenzoic acid,3-chloro-2-methybenzoic acid
82010
3-chloro-2-methylbenzoic acid
CC1=C(C=CC=C1Cl)C(=O)O
Packaging Glass Bottle
Quantity 1g
Infrared Spectrum Authentic
Melting Point 163.0°C to 166.0°C
Assay Percent Range 97.5% min. (HPLC)
Formula Weight 170.6
Percent Purity 98%
Chemical Name or Material 3-Chloro-2-methylbenzoic acid

GHS H Statement Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.

GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Call a POISON CENTER or doctor/physician if you feel unwell.

GHS Signal Word: Warning