Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-Chloro-2-methylpropene, 98%

Click to view available options
Quantity:
50g
250g
1000g
563-47-3
C4H7Cl
90.55
MFCD00000953
OHXAOPZTJOUYKM-UHFFFAOYSA-N
3-chloro-2-methylpropene,methallyl chloride,3-chloro-2-methyl-1-propene,1-propene, 3-chloro-2-methyl,2-methylallyl chloride,isobutenyl chloride,2-methyl-2-propenyl chloride,beta-methallyl chloride,propene, 3-chloro-2-methyl,cloruro di metallile
11241
CHEBI:82419
3-chloro-2-methylprop-1-ene
CC(=C)CCl
563-47-3
C4H7Cl
90.55
MFCD00000953
OHXAOPZTJOUYKM-UHFFFAOYSA-N
3-chloro-2-methylpropene,methallyl chloride,3-chloro-2-methyl-1-propene,1-propene, 3-chloro-2-methyl,2-methylallyl chloride,isobutenyl chloride,2-methyl-2-propenyl chloride,beta-methallyl chloride,propene, 3-chloro-2-methyl,cloruro di metallile
11241
CHEBI:82419
3-chloro-2-methylprop-1-ene
CC(=C)CCl
Applications
3-Chloro-2-methylpropene has been used as reactant in the synthesis of cyclobutanone. It is used as an intermediate in the production of plastics, pharmaceuticals and other organic chemicals.

Solubility
Not miscible in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Boiling Point 71°C to 72°C
Flash Point −12°C (9°F)
Melting Point -80°C
Quantity 50g
UN Number UN2554
Merck Index 14,2148
Formula Weight 90.55
Percent Purity 98%
Chemical Name or Material 3-Chloro-2-methylpropene

GHS H Statement
H225-H314-H302-H332-H317
Highly flammable liquid and vapour.
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful if inhaled.
May cause an allergic skin reaction.

GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: METHYL ALLYL CHLORIDE

RTECSNumber : UC8050000

TSCA : TSCA