Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Chloro-2-methylpropene (Tech.), 90%, ACROS Organics™

Click to view available options
Quantity:
50mL
250mL
1L
Packaging:
Glass bottle
563-47-3
C4H7Cl
90.55
MFCD00000953
OHXAOPZTJOUYKM-UHFFFAOYSA-N
3-chloro-2-methylpropene,methallyl chloride,3-chloro-2-methyl-1-propene,1-propene, 3-chloro-2-methyl,2-methylallyl chloride,isobutenyl chloride,2-methyl-2-propenyl chloride,beta-methallyl chloride,propene, 3-chloro-2-methyl,cloruro di metallile
11241
CHEBI:82419
3-chloro-2-methylprop-1-ene
CC(=C)CCl
563-47-3
C4H7Cl
90.55
MFCD00000953
OHXAOPZTJOUYKM-UHFFFAOYSA-N
3-chloro-2-methylpropene,methallyl chloride,3-chloro-2-methyl-1-propene,1-propene, 3-chloro-2-methyl,2-methylallyl chloride,isobutenyl chloride,2-methyl-2-propenyl chloride,beta-methallyl chloride,propene, 3-chloro-2-methyl,cloruro di metallile
11241
CHEBI:82419
3-chloro-2-methylprop-1-ene
CC(=C)CCl
Melting Point -80°C
Boiling Point 71°C to 72°C
Flash Point -18°C
Density 0.9200g/mL
Specific Gravity 0.92
Viscosity 0.42 mPa.s (20°C)
Refractive Index 1.4262 to 1.4282
Quantity 250mL
Water 0.1% max.
Infrared Spectrum Authentic
Color Undesignated
Packaging Glass bottle
CAS Min % 88.0
CAS Max % 100.0
Assay Percent Range 90%
Linear Formula H2C=C(CH3)CH2Cl
Beilstein 01, 209
Merck Index 15, 2150
Solubility Information Solubility in water: 0.5g/L (20°C). Other solubilities: miscible with most common organic solvents
Formula Weight 90.55
Physical Form Liquid
Percent Purity ≥88%
Grade Technical
Chemical Name or Material 3-Chloro-2-methylpropene, tech., 90%
Show More Show Less

GHS H Statement
Causes severe skin burns and eye damage. May cause an allergic skin reaction. Harmful if swallowed. Harmful if inhaled. Toxic to aquatic life with long lasting effects. Highly flammable liquid and vapour.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Avoid release to the environment. Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Ground/Bond container and receiving equipment.

Danger

EINECSNumber : 209-251-2

RTECSNumber : UC8050000

TSCA : TSCA