Alfa Aesar™ 3-Chloro-3-methyl-1-butyne, 95%

Manufacturer: Alfa Aesar™ B2525203

Quantity 1g

 View more versions of this product

Catalog No. AAB2525203

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1111-97-3
    Molecular Formula C5H7Cl
    Formula Weight 102.57
    MDL Number MFCD00190221
    UN Number UN1993
    Synonym 1,1-Dimethylpropargyl chloride
    Chemical Name or Material 3-Chloro-3-methyl-1-butyne

    Specifications

    Formula Weight 102.57
    Quantity 1g
    TSCA TSCA
    Odor Recognizable
    Molecular Formula C5H7Cl
    MDL Number MFCD00190221
    CAS 1111-97-3
    UN Number UN1993
    Synonym 1,1-Dimethylpropargyl chloride
    Health Hazard 2 GHS H Statement
    H225-H315-H319-H335
    Highly flammable liquid and vapour.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Recommended Storage 2° to 8°C
    DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
    Flash Point 4°C (40°F)
    Boiling Point 73° to 75°C
    Melting Point −61°C
    Health Hazard 1 Danger
    Health Hazard 3 GHS P Statement
    P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Chemical Name or Material 3-Chloro-3-methyl-1-butyne
    Assay 95%

    GHS H Statement
    H225-H315-H319-H335
    Highly flammable liquid and vapour.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    2° to 8°C

    GHS P Statement
    P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit