Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-chloro-3-methyl-1-butyne, 96%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass bottle
1111-97-3
C5H7Cl
102.56
MFCD00190221
QSILYWCNPOLKPN-UHFFFAOYSA-N
3-chloro-3-methyl-1-butyne,1-butyne, 3-chloro-3-methyl,3-chloro-3-methylbutyne,3-methyl-3-chlorobutyne,acmc-1bo5c,2-chloro-2-methyl butyne,3-chloro-3-methyl butyne,3-chloro-3-methyl-butyne,3-chloro-3-methyl-1-butyn,2-chloro-2-methyl-3-butyne
70679
3-chloro-3-methylbut-1-yne
CC(C)(C#C)Cl
1111-97-3
C5H7Cl
102.56
MFCD00190221
QSILYWCNPOLKPN-UHFFFAOYSA-N
3-chloro-3-methyl-1-butyne,1-butyne, 3-chloro-3-methyl,3-chloro-3-methylbutyne,3-methyl-3-chlorobutyne,acmc-1bo5c,2-chloro-2-methyl butyne,3-chloro-3-methyl butyne,3-chloro-3-methyl-butyne,3-chloro-3-methyl-1-butyn,2-chloro-2-methyl-3-butyne
70679
3-chloro-3-methylbut-1-yne
CC(C)(C#C)Cl
Boiling Point 73°C to 75°C
Flash Point 4°C
Specific Gravity 0.91
Density 0.9100g/mL
Refractive Index 1.4170 to 1.419
Infrared Spectrum Authentic
Quantity 1g
Color Colorless to Straw
Packaging Glass bottle
CAS Min % 95.0
CAS Max % 100.0
Assay Percent Range 95% min. (GC)
Formula Weight 102.56
Physical Form Liquid
Percent Purity 96%
Chemical Name or Material 3-Chloro-3-methyl-1-butyne
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.
Harmful in contact with skin.
Harmful if inhaled.
Highly flammable liquid and vapor.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 214-188-9