Alfa Aesar™ 3-Chloro-4-cyanopyridine, 95%

Manufacturer: Alfa Aesar™ H2718706

Quantity 5g

 View more versions of this product

Catalog No. AAH2718706

    N/A

Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-766-7000 or send an email to CS.WebEmail@thermofisher.com for assistance. 


    Chemical Identifiers

    CAS 68325-15-5
    Molecular Formula C6H3ClN2
    Formula Weight 138.56
    MDL Number MFCD05663706
    UN Number UN3439
    Synonym 3-Chloroisonicotinonitrile; 3-Chloropyridine-4-carbonitrile
    Chemical Name or Material 3-Chloro-4-cyanopyridine

    Specifications

    Formula Weight 138.56
    Molecular Formula C6H3ClN2
    MDL Number MFCD05663706
    UN Number UN3439
    Synonym 3-Chloroisonicotinonitrile; 3-Chloropyridine-4-carbonitrile
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H311-H302-H332-H315-H319-H335
    Toxic in contact with skin.
    Harmful if swallowed.
    Harmful if inhaled.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P280h-P305+P351+P338-P309-P310
    Wear protective gloves/clothing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
    Melting Point 71° to 74°C
    Quantity 5g
    CAS 68325-15-5
    Chemical Name or Material 3-Chloro-4-cyanopyridine
    Assay 95%

    GHS H Statement
    H311-H302-H332-H315-H319-H335
    Toxic in contact with skin.
    Harmful if swallowed.
    Harmful if inhaled.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P280h-P305+P351+P338-P309-P310
    Wear protective gloves/clothing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit