Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3'-Chloro-4'-fluoroacetanilide, 98%

Click to view available options
Quantity:
5g
25g
100g
877-90-7
C8H7ClFNO
187.598
MFCD00018095
ALPHMTFVUKDBGJ-UHFFFAOYSA-N
n-3-chloro-4-fluorophenyl acetamide,3'-chloro-4'-fluoroacetanilide,3-chloro-4-fluoroacetanilide,3-chloro-4-fluoroacetanililide,n-acetyl-3-chloro-4-fluoroaniline,3'-chloro-4'-fluoro acetanilide,acetamide,n-3-chloro-4-fluorophenyl,n-3-chloranyl-4-fluoranyl-phenyl ethanamide
589188
N-(3-chloro-4-fluorophenyl)acetamide
CC(=O)NC1=CC(=C(C=C1)F)Cl
877-90-7
C8H7ClFNO
187.598
MFCD00018095
ALPHMTFVUKDBGJ-UHFFFAOYSA-N
n-3-chloro-4-fluorophenyl acetamide,3'-chloro-4'-fluoroacetanilide,3-chloro-4-fluoroacetanilide,3-chloro-4-fluoroacetanililide,n-acetyl-3-chloro-4-fluoroaniline,3'-chloro-4'-fluoro acetanilide,acetamide,n-3-chloro-4-fluorophenyl,n-3-chloranyl-4-fluoranyl-phenyl ethanamide
589188
N-(3-chloro-4-fluorophenyl)acetamide
CC(=O)NC1=CC(=C(C=C1)F)Cl
Melting Point 116°C to 119°C
Quantity 25g
Formula Weight 187.6
Percent Purity 98%
Chemical Name or Material 3'-Chloro-4'-fluoroacetanilide

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning