Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-Chloro-4-fluoroaniline, 98+%

Manufacturer:  Alfa Aesar™ A1185822

Quantity 100g

 View more versions of this product

Catalog No. AAA1185822


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

367-21-5
145.56
UN2811
3-Chloro-4-fluoroaniline
C6H5ClFN
MFCD00007767
1562786

Specifications

149°C (300°F)
44° to 47°C
367-21-5
C6H5ClFN
MFCD00007767
1562786
≥98%
227° to 230°C
100g
Characteristic
145.56
UN2811
3-Chloro-4-fluoroaniline

Safety and Handling

SDS & Safety Documents

GHS H Statement
H311-H373-H302-H332-H315-H319
Toxic in contact with skin.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P260-P261-P305+P351+P338-P361-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit