Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Chloro-4-fluoroaniline, 98%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
Packaging:
Glass bottle
367-21-5
C6H5ClFN
145.56
MFCD00007767
YSEMCVGMNUUNRK-UHFFFAOYSA-N
benzenamine, 3-chloro-4-fluoro, 3-chloro-4-fluorobenzenamine, 3-chloro-4-fluoro-phenylamine, aniline, 3-chloro-4-fluoro, 3-chloro-4-fluoro aniline, unii-7l63cc70uq, 3-chloro-4-fluorophenylamine, 4-fluoro-3-chloroaniline, 3-chloro-4-fluoro-aniline, 3-chlor-4-fluoranilin
9708
3-chloro-4-fluoroaniline
NC1=CC=C(F)C(Cl)=C1
367-21-5
C6H5ClFN
145.56
MFCD00007767
YSEMCVGMNUUNRK-UHFFFAOYSA-N
benzenamine, 3-chloro-4-fluoro, 3-chloro-4-fluorobenzenamine, 3-chloro-4-fluoro-phenylamine, aniline, 3-chloro-4-fluoro, 3-chloro-4-fluoro aniline, unii-7l63cc70uq, 3-chloro-4-fluorophenylamine, 4-fluoro-3-chloroaniline, 3-chloro-4-fluoro-aniline, 3-chlor-4-fluoranilin
9708
3-chloro-4-fluoroaniline
NC1=CC=C(F)C(Cl)=C1
Boiling Point 230°C
Flash Point 149°C
Quantity 100g
Infrared Spectrum Authentic
Packaging Glass bottle
Melting Point 42°C to 47°C
Color Beige to Brown
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula ClC6H3(F)NH2
Beilstein 12, I, 309
Solubility Information Solubility in water: 10g/L water 520°C). Other solubilities: soluble in organic solvents
Formula Weight 145.56
Physical Form Crystalline Solid
Percent Purity 98%
Chemical Name or Material 3-Chloro-4-fluoroaniline, 98%
Show More Show Less

GHS H Statement
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Harmful if inhaled.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor

GHS Signal Word: Warning

EINECSNumber : 206-682-8

TSCA : TSCA

Recommended Storage : Product darkens on prolonged storage