Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-Chloro-4-fluorobenzeneboronic acid, 98%

Click to view available options
Quantity:
1g
5g
25g
144432-85-9
C6H5BClFO2
174.362
MFCD00051800
WJDZZXIDQYKVDG-UHFFFAOYSA-N
3-chloro-4-fluorobenzeneboronic acid,3-chloro-4-fluorophenyl boronic acid,3-chloro-4-fluorophenyl boranediol,4-fluoro-3-chlorophenylboronic acid,3-chloro-4-fluoro phenyl boronic acid,3-chloro-4-fluoro-phenyl boronic acid,boronic acid, 3-chloro-4-fluorophenyl,b-3-chloro-4-fluorophenyl-boronic acid,pubchem1780
2734660
(3-chloro-4-fluorophenyl)boronic acid
B(C1=CC(=C(C=C1)F)Cl)(O)O
144432-85-9
C6H5BClFO2
174.362
MFCD00051800
WJDZZXIDQYKVDG-UHFFFAOYSA-N
3-chloro-4-fluorobenzeneboronic acid,3-chloro-4-fluorophenyl boronic acid,3-chloro-4-fluorophenyl boranediol,4-fluoro-3-chlorophenylboronic acid,3-chloro-4-fluoro phenyl boronic acid,3-chloro-4-fluoro-phenyl boronic acid,boronic acid, 3-chloro-4-fluorophenyl,b-3-chloro-4-fluorophenyl-boronic acid,pubchem1780
2734660
(3-chloro-4-fluorophenyl)boronic acid
B(C1=CC(=C(C=C1)F)Cl)(O)O
Applications
3-Chloro-4-fluorobenzeneboronic acid is used as a reactant for Rh-catalyzed asymmetric addition reactions, palladium-catalyzed oxidative cross-coupling reaction and Suzuki-Miyaura coupling.

Notes
Incompatible with strong oxidizing agents.
Melting Point 255°C to 260°C
Quantity 25g
Beilstein 7369789
Formula Weight 174.37
Percent Purity 98%
Chemical Name or Material 3-Chloro-4-fluorobenzeneboronic acid

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning