Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-Chloro-4-fluorobenzonitrile, 98+%

Click to view available options
Quantity:
5g
25g
100g
117482-84-5
C7H3ClFN
155.556
MFCD00015431
VAHXXQJJZKBZDX-UHFFFAOYSA-N
3-chloro-4-fluoro-benzonitrile,benzonitrile, 3-chloro-4-fluoro,3-chloro-4-fluorobenzenecarbonitrile,pubchem1561,acmc-1bnmf,3-chloro-4-florobenzonitrile,3-chloro-4-fluorobenzontrile,ksc174m4n,3-chloro,4-fluorobenzonitrile,3-chloro4-fluorobenzo-nitrile
145525
3-chloro-4-fluorobenzonitrile
C1=CC(=C(C=C1C#N)Cl)F
117482-84-5
C7H3ClFN
155.556
MFCD00015431
VAHXXQJJZKBZDX-UHFFFAOYSA-N
3-chloro-4-fluoro-benzonitrile,benzonitrile, 3-chloro-4-fluoro,3-chloro-4-fluorobenzenecarbonitrile,pubchem1561,acmc-1bnmf,3-chloro-4-florobenzonitrile,3-chloro-4-fluorobenzontrile,ksc174m4n,3-chloro,4-fluorobenzonitrile,3-chloro4-fluorobenzo-nitrile
145525
3-chloro-4-fluorobenzonitrile
C1=CC(=C(C=C1C#N)Cl)F
Quantity 100g
Melting Point 68°C to 72°C
UN Number UN3439
Beilstein 4740501
Formula Weight 155.56
Percent Purity ≥98%
Chemical Name or Material 3-Chloro-4-fluorobenzonitrile

GHS H Statement
H311-H302-H332-H315-H319
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.

EINECSNumber : 000-000-0

TSCA : No

Recommended Storage : Ambient temperatures