Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alfa Aesar™ 3-Chloro-4-fluorobenzyl alcohol, 97%
Click to view available options
Quantity:
1g
5g
Description
Applications
3-Chloro-4-fluorobenzyl alcohol is used as a pharmaceutical intermediate.
Solubility
Slightly soluble in water.
Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
3-Chloro-4-fluorobenzyl alcohol is used as a pharmaceutical intermediate.
Solubility
Slightly soluble in water.
Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
Specifications
Refractive Index | 1.5355 |
Quantity | 5g |
Solubility Information | Slightly soluble in water. |
Formula Weight | 160.58 |
Percent Purity | 97% |
Chemical Name or Material | 3-Chloro-4-fluorobenzyl alcohol |
Safety and Handling
GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
H315-H319
TSCA : No
Recommended Storage : Ambient temperatures