Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-Chloro-4'-fluoropropiophenone, 97%

Click to view available options
Quantity:
5g
25g
100g
347-93-3
C9H8ClFO
186.61
MFCD00000991
AAHQPLJUSLMHHR-UHFFFAOYSA-N
3-chloro-4'-fluoropropiophenone,3-chloro-1-4-fluorophenyl propan-1-one,3-chloro-1-4-fluorophenyl-1-propanone,1-propanone, 3-chloro-1-4-fluorophenyl,p-fluoro-,a-chloropropiophenone,pubchem13250,acmc-1ctxg,ksc573a4j,3-chloro-4'fluoropropiophenone,3-chloro4'-fluoropropiophenone
67672
3-chloro-1-(4-fluorophenyl)propan-1-one
C1=CC(=CC=C1C(=O)CCCl)F
347-93-3
C9H8ClFO
186.61
MFCD00000991
AAHQPLJUSLMHHR-UHFFFAOYSA-N
3-chloro-4'-fluoropropiophenone,3-chloro-1-4-fluorophenyl propan-1-one,3-chloro-1-4-fluorophenyl-1-propanone,1-propanone, 3-chloro-1-4-fluorophenyl,p-fluoro-,a-chloropropiophenone,pubchem13250,acmc-1ctxg,ksc573a4j,3-chloro-4'fluoropropiophenone,3-chloro4'-fluoropropiophenone
67672
3-chloro-1-(4-fluorophenyl)propan-1-one
C1=CC(=CC=C1C(=O)CCCl)F
Quantity 5g
Boiling Point 100°C to 103°C (3mmHg)
Melting Point 46°C to 49°C
Beilstein 1866785
Formula Weight 186.61
Percent Purity 97%
Chemical Name or Material 3-Chloro-4'-fluoropropiophenone

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning