Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-Chloro-4-hydroxy-5-methoxybenzyl alcohol, 98%

Click to view available options
Quantity:
1g
5g
25g
20624-92-4
C8H9ClO3
188.607
MFCD00016870
VQEZBFOBPUKDGM-UHFFFAOYSA-N
5-chlorovanillyl alcohol,2-chloro-4-hydroxymethyl-6-methoxyphenol,3-chloro-4-hydroxy-5-methoxybenzyl alcohol,3-chloro-4-hydroxy-5-methoxybenzenemethanol,acmc-20am3y,6-chloro-4-hydroxymethyl-2-methoxyphenol,5-chloro-4-hydroxy-3-methoxybenzenemethanol,benzenemethanol,3-chloro-4-hydroxy-5-methoxy
4181655
2-chloro-4-(hydroxymethyl)-6-methoxyphenol
COC1=C(C(=CC(=C1)CO)Cl)O
20624-92-4
C8H9ClO3
188.607
MFCD00016870
VQEZBFOBPUKDGM-UHFFFAOYSA-N
5-chlorovanillyl alcohol,2-chloro-4-hydroxymethyl-6-methoxyphenol,3-chloro-4-hydroxy-5-methoxybenzyl alcohol,3-chloro-4-hydroxy-5-methoxybenzenemethanol,acmc-20am3y,6-chloro-4-hydroxymethyl-2-methoxyphenol,5-chloro-4-hydroxy-3-methoxybenzenemethanol,benzenemethanol,3-chloro-4-hydroxy-5-methoxy
4181655
2-chloro-4-(hydroxymethyl)-6-methoxyphenol
COC1=C(C(=CC(=C1)CO)Cl)O
Melting Point 115°C to 118°C
Quantity 25g
Formula Weight 188.62
Percent Purity 98%
Chemical Name or Material 3-Chloro-4-hydroxy-5-methoxybenzyl alcohol

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning