Alfa Aesar™ 3'-Chloro-4'-methoxyacetophenone, 96%

Manufacturer: Alfa Aesar™ H2582603

Quantity 1g

 View more versions of this product

Catalog No. AAH2582603

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 37612-52-5
    Molecular Formula C9H9ClO2
    Formula Weight 184.62
    MDL Number MFCD04973413
    Synonym 1-(3-Chloro-4-methoxyphenyl)ethanone
    Chemical Name or Material 3'-Chloro-4'-methoxyacetophenone

    Specifications

    Formula Weight 184.62
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Melting Point 74° to 76°C
    Quantity 1g
    Molecular Formula C9H9ClO2
    MDL Number MFCD04973413
    CAS 37612-52-5
    Synonym 1-(3-Chloro-4-methoxyphenyl)ethanone
    Health Hazard 3 GHS P Statement
    P280g-P305+P351+P338
    Wear protective gloves.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Chemical Name or Material 3'-Chloro-4'-methoxyacetophenone
    Assay 96%

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P280g-P305+P351+P338
    Wear protective gloves.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit