Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Chloro-4-methoxybenzoic acid, 98+%, Thermo Scientific Chemicals

Catalog Number p-7080540
Click to view available options
Quantity:
5 g
25 g
37908-96-6
C8H7ClO3
186.591
MFCD00016512
IBCQUQXCTOPJOD-UHFFFAOYSA-N
3-chloro-p-anisic acid, 3-chloro-4-methoxybenzolic acid, benzoic acid, 3-chloro-4-methoxy, pubchem19130, acmc-209iw4, ksc494s0j, 3-chloro-4-methoxybenzoicacid, rarechem al bo 2400, 3-chloro-4-anisic acid, 3-chloro-4-methoxy benzoic acid
169982
3-chloro-4-methoxybenzoic acid
COC1=C(C=C(C=C1)C(=O)O)Cl
37908-96-6
C8H7ClO3
186.591
MFCD00016512
IBCQUQXCTOPJOD-UHFFFAOYSA-N
3-chloro-p-anisic acid, 3-chloro-4-methoxybenzolic acid, benzoic acid, 3-chloro-4-methoxy, pubchem19130, acmc-209iw4, ksc494s0j, 3-chloro-4-methoxybenzoicacid, rarechem al bo 2400, 3-chloro-4-anisic acid, 3-chloro-4-methoxy benzoic acid
169982
3-chloro-4-methoxybenzoic acid
COC1=C(C=C(C=C1)C(=O)O)Cl

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 215°C to 219°C
Quantity 5 g
Beilstein 2087584
Formula Weight 186.59
Percent Purity ≥98%
Chemical Name or Material 3-Chloro-4-methoxybenzoic acid
Hazard Category H301-H319
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P264b-P270-P280i-P301+P310-P305+P351+P338-P330-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 253-708-9
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit