Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
3-Chloro-4-methoxybenzoic Acid 97.0+%, TCI America™
Click to view available options
Quantity:
5 g
25 g
Specifications
Melting Point | 217°C |
Quantity | 25 g |
Formula Weight | 186.59 |
Percent Purity | ≥97.0% (GC,T) |
Physical Form | Crystalline Powder |
Chemical Name or Material | 3-Chloro-4-methoxybenzoic Acid |
Safety and Handling
TSCA : No